3-(Hydroxymethyl)phenylboronic acid pinacol ester ≥98%
Supplier: Matrix Scientific
Synonyms:
2-[3-(Hydroxymethyl)phenyl]-4,4,5,5-tetramethyl-1,3,2-dioxaborolane, 3-Hydroxymethylphenylboronic acid pinacol ester, 3-(4,4,5,5-Tetramethyl-1,3,2-dioxaborolan-2-yl)benzyl alcohol, (3-(4,4,5,5-Tetramethyl-1,3,2-dioxaborolan-2-yl)phenyl)methanol, 3-(Hydroxymethyl)phenylboronic acid Pinacol ester, 3-(Hydroxymethyl)benzeneboronic acid pinacol ester
069746-1G
069746-5G
069746-25G
101941-738EA
146.15
USD
101941-738
101941-740
101941-742
3-(Hydroxymethyl)phenylboronic acid pinacol ester ≥98%
3-(Hydroxymethyl)phenylboronic acid pinacol ester
Hazard: Irritant
Formula:
C₁₃H₁₉BO₃ MW: 234.10 g/mol |
MDL Number:
MFCD09266196 CAS Number: 443776-76-9 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...