5-Bromothiazole-4-carboxylic acid

Supplier: Matrix Scientific

069745-1G 069745-5G
101941-734EA 196.92 USD
101941-734 101941-736
5-Bromothiazole-4-carboxylic acid
5-Bromothiazole-4-carboxylic acid

Hazard: Irritant

Formula: C₄H₂BrNO₂S
MW: 208.04 g/mol
Melting Pt: 182…187 °C
MDL Number: MFCD08275714
CAS Number: 103878-58-6

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR