Methyl-5-bromothiazole-4-carboxylate ≥98%

Supplier: Matrix Scientific

Synonyms: Methyl 5-bromothiazole-4-carboxylate

069742-1G 069742-5G
101941-722EA 121.54 USD
101941-722 101941-724
Methyl-5-bromothiazole-4-carboxylate ≥98%
Methyl-5-bromothiazole-4-carboxylate

Hazard: Irritant

Formula: C₅H₄BrNO₂S
MW: 222.07 g/mol
Melting Pt: 98…101 °C
MDL Number: MFCD08275713
CAS Number: 913836-22-3

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR