6-Aminopyridin-2-ol ≥98%

Supplier: Matrix Scientific

Synonyms: 6-Hydroxypyridin-2-amine, 6-Aminopyridin-2-ol

069741-1G 069741-5G 069741-25G
101941-716EA 115.38 USD
101941-716 101941-718 102551-756
6-Aminopyridin-2-ol ≥98%
6-Aminopyridin-2-ol

Hazard: Irritant

Formula: C₅H₆N₂O
MW: 110.11 g/mol
MDL Number: MFCD00039717
CAS Number: 59315-47-8

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR