6-Hydroxypyrimidine-4-carboxylic acid ≥97%

Supplier: Matrix Scientific

069740-5G 069740-25G
101941-712EA 113.85 USD
101941-712 101941-714
6-Hydroxypyrimidine-4-carboxylic acid ≥97%
6-Hydroxypyrimidine-4-carboxylic acid

Hazard: Irritant

Formula: C₅H₄N₂O₃
MDL Number: MFCD00023267
CAS Number: 6299-87-2

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR