4-Fluoro-3,5-bis(tributylstannyl)pyridine ≥97%
Supplier: Matrix Scientific
069739-1G
069739-5G
101941-706EA
815.38
USD
101941-706
101941-708
4-Fluoro-3,5-bis(tributylstannyl)pyridine ≥97%
4-Fluoro-3,5-bis(tributylstannyl)pyridine
Hazard: Irritant
Formula:
C₂₉H₅₆FNSn₂ |
MDL Number:
MFCD10699174 CAS Number: 1204580-75-5 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...