3-Fluoro-4-nitropyridine ≥96%

Supplier: Matrix Scientific

069736-1G 069736-5G
101941-692EA 392.31 USD
101941-692 101941-694
3-Fluoro-4-nitropyridine ≥96%
3-Fluoro-4-nitropyridine

Hazard: Toxic/Refrigerate

Formula: C₅H₃FN₂O₂
Boiling Pt: 62-64°C/5mmHg
MDL Number: MFCD04112517
CAS Number: 13505-01-6

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR