6-Amino-1-(tetrahydropyranyl)-1H-indazole ≥98%

Supplier: Matrix Scientific

Synonyms: 1-(Tetrahydro-2H-pyran-2-yl)-1H-indazol-6-amine, 6-Amino-1-(tetrahydro-2H-pyran-2-yl)-1H-indazole

069734-500MG 069734-1G 069734-5G
101941-680EA 698.46 USD
101941-680 101941-682 101941-684
6-Amino-1-(tetrahydropyranyl)-1H-indazole ≥98%
6-Amino-1-(tetrahydropyranyl)-1H-indazole

Hazard: Irritant

Formula: C₁₂H₁₅N₃O
MW: 217.27 g/mol
MDL Number: MFCD10699171
CAS Number: 1053655-59-6

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR