5-Amino-1-(tetrahydropyranyl)-1H-indazole ≥98%

Supplier: Matrix Scientific

Synonyms: 1-(Tetrahydro-2H-pyran-2-yl)-1H-indazol-5-amine

069733-500MG 069733-1G 069733-5G
101941-674EA 263.08 USD
101941-674 101941-676 101941-678
5-Amino-1-(tetrahydropyranyl)-1H-indazole ≥98%
5-Amino-1-(tetrahydropyranyl)-1H-indazole

Hazard: Irritant

Formula: C₁₂H₁₅N₃O
MW: 217.27 g/mol
MDL Number: MFCD10699170
CAS Number: 478832-10-9

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR