4-Amino-1-(tetrahydropyranyl)-1H-indazole ≥97%

Supplier: Matrix Scientific

Synonyms: 1-(Tetrahydro-2H-pyran-2-yl)-1H-indazol-4-amine

069731-1G 069731-5G 069731-25G
101941-662EA 233.85 USD
101941-662 101941-664 101941-666
4-Amino-1-(tetrahydropyranyl)-1H-indazole ≥97%
4-Amino-1-(tetrahydropyranyl)-1H-indazole

Hazard: Irritant

Formula: C₁₂H₁₅N₃O
MW: 217.27 g/mol
MDL Number: MFCD10699168
CAS Number: 1053655-57-4

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR