2-(Tributylstannyl)propene ≥97%

Supplier: Matrix Scientific

069729-1G 069729-5G 069729-25G
101941-650EA 230.77 USD
101941-650 101941-652 101941-654
2-(Tributylstannyl)propene ≥97%
2-(Tributylstannyl)propene

Hazard: Irritant

Formula: C₁₅H₃₂Sn
MDL Number: MFCD10699166
CAS Number: 100073-15-2

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR