2,3-Dihydrobenzo[b][1,4]dioxine-5-carboxylic acid ≥98%

Supplier: Matrix Scientific

Synonyms: 2,3-dihydro-1,4-benzodioxine-5-carboxylic acid, 2,3-dihydrobenzo[b][1,4]dioxine-5-carboxylic acid, 1,4-Benzodioxan-5-carboxylic acid

069727-1G 069727-5G
101941-640EA 15.38 USD
101941-640 101941-642
2,3-Dihydrobenzo[b][1,4]dioxine-5-carboxylic acid ≥98%
2,3-Dihydrobenzo[b][1,4]dioxine-5-carboxylic acid

Hazard: Irritant

Formula: C₉H₈O₄
MW: 180.16 g/mol
MDL Number: MFCD00239415
CAS Number: 4442-53-9

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR