5-Methyloxazole-4-carboxylic acid ≥98%

Supplier: Matrix Scientific

Synonyms: 4-Oxazolecarboxylicacid, 5-methyl-, 5-Methyl-1,3-oxazole-4-carboxylic acid, 4-Carboxy-5-methyl-1,3-oxazole

069725-1G 069725-5G
101941-632EA 228.85 USD
101941-632 101941-634
5-Methyloxazole-4-carboxylic acid ≥98%
5-Methyloxazole-4-carboxylic acid

Hazard: Irritant

Formula: C₅H₅NO₃
MW: 127.1 g/mol
MDL Number: MFCD08056309
CAS Number: 103879-58-9

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR