7-Nitroso-5,6,7,8-tetrahydroimidazo[1,2-a]pyrazine

Supplier: Matrix Scientific

069724-500MG 069724-1G
101941-626EA 815.38 USD
101941-626 101941-628
7-Nitroso-5,6,7,8-tetrahydroimidazo[1,2-a]pyrazine
7-Nitroso-5,6,7,8-tetrahydroimidazo[1,2-a]pyrazine

Hazard: Irritant

Formula: C₆H₈N₄O
MW: 152.16 g/mol
MDL Number: MFCD10699164
CAS Number: 91476-88-9

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR