5-Methoxy-3-(tributylstannyl)pyridine ≥96%
Supplier: Matrix Scientific
069719-1G
069719-5G
101941-598EA
349.23
USD
101941-598
101941-600
5-Methoxy-3-(tributylstannyl)pyridine ≥96%
5-Methoxy-3-(tributylstannyl)pyridine
Hazard: Irritant
Formula:
C₁₈H₃₃NOSn |
MDL Number:
MFCD10699160 CAS Number: 1204580-74-4 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...