3-Methoxy-2-(tributylstannyl)pyridine ≥94%

Supplier: Matrix Scientific

069718-1G 069718-5G
101941-594EA 524.62 USD
101941-594 101941-596
3-Methoxy-2-(tributylstannyl)pyridine ≥94%
3-Methoxy-2-(tributylstannyl)pyridine

Hazard: Irritant

Formula: C₁₈H₃₃NOSn
MDL Number: MFCD10699159
CAS Number: 1094072-15-7

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR