6-Chloro-3-(tributylstannyl)pyridine ≥97%

Supplier: Matrix Scientific

069717-1G 069717-5G
101941-590EA 349.23 USD
101941-590 101941-592
6-Chloro-3-(tributylstannyl)pyridine ≥97%
6-Chloro-3-(tributylstannyl)pyridine

Hazard: Irritant

Formula: C₁₇H₃₀ClNSn
MDL Number: MFCD01318969
CAS Number: 183545-05-3

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR