5-Chloro-3-(tributylstannyl)pyridine ≥92%
Supplier: Matrix Scientific
069716-1G
069716-5G
101941-586EA
407.69
USD
101941-586
101941-588
5-Chloro-3-(tributylstannyl)pyridine ≥92%
5-Chloro-3-(tributylstannyl)pyridine
Hazard: Irritant
Formula:
C₁₇H₃₀ClNSn |
MDL Number:
MFCD10699158 CAS Number: 206115-67-5 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...