3-Chloro-2-(tributylstannyl)pyridine ≥97%

Supplier: Matrix Scientific

069715-1G 069715-5G
101941-582EA 640 USD
101941-582 101941-584
3-Chloro-2-(tributylstannyl)pyridine ≥97%
3-Chloro-2-(tributylstannyl)pyridine

Hazard: Irritant

Formula: C₁₇H₃₀ClNSn
MDL Number: MFCD10699157
CAS Number: 206357-78-0

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR