5-Chloro-2-(tributylstannyl)pyridine ≥96%

Supplier: Matrix Scientific

069714-1G 069714-5G
101941-578EA 276.92 USD
101941-578 101941-580
5-Chloro-2-(tributylstannyl)pyridine ≥96%
5-Chloro-2-(tributylstannyl)pyridine

Hazard: Irritant

Formula: C₁₇H₃₀ClNSn
MDL Number: MFCD10699156
CAS Number: 611168-63-9

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR