2-Chloro-4-(tributylstannyl)pyridine ≥97%

Supplier: Matrix Scientific

069712-1G 069712-5G
101941-570EA 276.92 USD
101941-570 101941-572
2-Chloro-4-(tributylstannyl)pyridine ≥97%
2-Chloro-4-(tributylstannyl)pyridine

Hazard: Irritant

Formula: C₁₇H₃₀ClNSn
MDL Number: MFCD10699154
CAS Number: 1204580-73-3

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR