2-Methoxy-4-(tributylstannyl)pyridine ≥95%

Supplier: Matrix Scientific

069711-1G 069711-5G
101941-566EA 640 USD
101941-566 101941-568
2-Methoxy-4-(tributylstannyl)pyridine ≥95%
2-Methoxy-4-(tributylstannyl)pyridine

Hazard: Irritant

Formula: C₁₈H₃₃NOSn
MDL Number: MFCD10699153
CAS Number: 1204580-72-2

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR