6-Methoxy-3-(tributylstannyl)pyridine ≥95%
Supplier: Matrix Scientific
069709-1G
069709-5G
101941-556EA
212.31
USD
101941-556
101941-558
6-Methoxy-3-(tributylstannyl)pyridine ≥95%
6-Methoxy-3-(tributylstannyl)pyridine
Hazard: Irritant
Formula:
C₁₈H₃₃NOSn |
MDL Number:
MFCD01319027 CAS Number: 164014-93-1 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...