2,3-Difluoro-4-(tributylstannyl)pyridine ≥95%

Supplier: Matrix Scientific

069708-1G 069708-5G
101941-552EA 930.77 USD
101941-552 101941-554
2,3-Difluoro-4-(tributylstannyl)pyridine ≥95%
2,3-Difluoro-4-(tributylstannyl)pyridine

Hazard: Irritant

Formula: C₁₇H₂₉F₂NSn
MDL Number: MFCD10699151
CAS Number: 1245816-05-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR