2-(Tributylstannyl)-1-tritylimidazole ≥95%

Supplier: Matrix Scientific

069707-500MG 069707-1G 069707-5G
101941-546EA 930.77 USD
101941-546 101941-548 101941-550
2-(Tributylstannyl)-1-tritylimidazole ≥95%
2-(Tributylstannyl)-1-tritylimidazole

Hazard: Irritant

Formula: C₃₄H₄₄N₂Sn
MDL Number: MFCD10699150
CAS Number: 208934-35-4

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR