1-(Tributylstannyl)-3-(trifluoromethyl)-1-propyne ≥90%

Supplier: Matrix Scientific

069703-1G 069703-5G
101941-528EA 792.31 USD
101941-528 101941-530
1-(Tributylstannyl)-3-(trifluoromethyl)-1-propyne ≥90%
1-(Tributylstannyl)-3-(trifluoromethyl)-1-propyne

Hazard: Irritant

Formula: C₁₆H₂₉F₃Sn
MDL Number: MFCD11109352
CAS Number: 64185-12-2

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR