Ethyl-2-chlorothiazole-4-acetate ≥96%

Supplier: Matrix Scientific

069702-1G 069702-5G
101941-524EA 698.46 USD
101941-524 101941-526
Ethyl-2-chlorothiazole-4-acetate ≥96%
Ethyl-2-chlorothiazole-4-acetate

Hazard: Irritant

Formula: C₇H₈ClNO₂S
MDL Number: MFCD10699148
CAS Number: 464185-24-8

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR