4-(Aminomethyl)phenylacetic acid ethyl ester hydrochloride ≥97%

Supplier: Matrix Scientific

Synonyms: Ethyl 2-(4-(aminomethyl)phenyl)acetate hydrochloride, Ethyl 4-(aminomethyl)phenylacetate hydrochloride

069701-250MG 069701-1G
101941-518EA 295.59 USD
101941-518 101941-522
4-(Aminomethyl)phenylacetic acid ethyl ester hydrochloride ≥97%
4-(Aminomethyl)phenylacetic acid ethyl ester hydrochloride

Hazard: Irritant

Formula: C₁₁H₁₆ClNO₂
MW: 229.71 g/mol
MDL Number: MFCD09953123
CAS Number: 17841-69-9

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR