1-(N-Boc)-3-cyanoazetidine ≥98%

Supplier: Matrix Scientific

Synonyms: tert-Butyl 3-cyanoazetidine-1-carboxylate, Azetidine-3-carbonitrile, N-BOC protected

069700-1G 069700-5G
101941-514EA 58.46 USD
101941-514 101941-516
1-(N-Boc)-3-cyanoazetidine ≥98%
1-(N-Boc)-3-cyanoazetidine

Hazard: Irritant

Formula: C₉H₁₄N₂O₂
MDL Number: MFCD06796640
CAS Number: 142253-54-1

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR