2-Chloro-1-(tert-butyl)-1H-imidazole
Supplier: Matrix Scientific
069699-500MG
069699-1G
069699-5G
101941-506EA
727.69
USD
101941-506
101941-508
101941-510
2-Chloro-1-(tert-butyl)-1H-imidazole
2-Chloro-1-(tert-butyl)-1H-imidazole
Hazard: Irritant
Formula:
C₇H₁₁ClN₂ MW: 158.63 g/mol |
MDL Number:
MFCD10699140 CAS Number: 1053655-54-1 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...