2,5-Dibromo-3-chloropyridine ≥97%

Supplier: Matrix Scientific

Synonyms: 3-Chloro-2,5-dibromopyridine

069695-1G 069695-5G
101941-486EA 24.62 USD
101941-486 101941-488
2,5-Dibromo-3-chloropyridine ≥97%
2,5-Dibromo-3-chloropyridine

Hazard: Irritant

Formula: C₅H₂Br₂ClN
MW: 271.3371 g/mol
MDL Number: MFCD08277306
CAS Number: 160599-70-2

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR