Thieno[2,3-c]pyridine ≥97%

Supplier: Matrix Scientific

069692-1G 069692-5G
101941-474EA 784.62 USD
101941-474 101941-476
Thieno[2,3-c]pyridine ≥97%
Thieno[2,3-c]pyridine

Hazard: Irritant

Formula: C₇H₅NS
MDL Number: MFCD01760492
CAS Number: 272-12-8

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR