Thiazol-2-ylmethanol ≥96%

Supplier: Matrix Scientific

Synonyms: 2-(Hydroxymethyl)thiazole, 2-Thiazolemethanol

069690-1G 069690-5G
101941-462EA 135.38 USD
101941-462 101941-464
Thiazol-2-ylmethanol ≥96%
Thiazol-2-ylmethanol

Hazard: Irritant/Refrigerate

Formula: C₄H₅NOS
MDL Number: MFCD06200855
CAS Number: 14542-12-2

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR