1-(Benzyloxy)-4-bromo-2,3-difluorobenzene ≥96%

Supplier: Matrix Scientific

069687-1G 069687-5G 069687-25G
101941-444EA 116.92 USD
101941-444 101941-446 101941-448
1-(Benzyloxy)-4-bromo-2,3-difluorobenzene ≥96%
1-(Benzyloxy)-4-bromo-2,3-difluorobenzene

Hazard: Irritant

Formula: C₁₃H₉BrF₂O
MDL Number: MFCD09475823
CAS Number: 941294-52-6

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR