5-Iodopyridine-2-carbonitrile ≥98%

Supplier: Matrix Scientific

Synonyms: 2-Cyano-5-iodopyridine

069686-1G 069686-5G 069686-25G
101941-438EA 524.62 USD
101941-438 101941-440 101941-442
5-Iodopyridine-2-carbonitrile ≥98%
5-Iodopyridine-2-carbonitrile

Hazard: Irritant

Formula: C₆H₃IN₂
MW: 230.0100 g/mol
MDL Number: MFCD10699132
CAS Number: 41960-47-8

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR