2,2-Dimethylethenylboronic acid ≥97%
Supplier: Matrix Scientific
Synonyms:
(2-Methylprop-1-en-1-yl)boronic acid
069685-1G
069685-5G
069685-25G
101941-432EA
209.23
USD
101941-432
101941-434
101941-436
2,2-Dimethylethenylboronic acid ≥97%
2,2-Dimethylethenylboronic acid
Hazard: Irritant
Formula:
C₄H₉BO₂ MW: 99.93 g/mol |
MDL Number:
MFCD01074609 CAS Number: 14559-88-7 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...