2,3-Difluoro-6-benzyloxyphenylboronic acid pinacol ester ≥98%

Supplier: Matrix Scientific

069683-500MG 069683-1G
101941-424EA 698.46 USD
101941-424 101941-426
2,3-Difluoro-6-benzyloxyphenylboronic acid pinacol ester ≥98%
2,3-Difluoro-6-benzyloxyphenylboronic acid pinacol ester

Hazard: Irritant

Formula: C₁₉H₂₁BF₂O₃
MDL Number: MFCD11109351
CAS Number: 1204580-70-0

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR