Methyl-2-chloro-4-thiazolecarboxylate ≥97%

Supplier: Matrix Scientific

069680-1G 069680-5G 069680-25G
101941-410EA 190.77 USD
101941-410 101941-412 101941-414
Methyl-2-chloro-4-thiazolecarboxylate ≥97%
Methyl-2-chloro-4-thiazolecarboxylate

Hazard: Irritant

Formula: C₅H₄ClNO₂S
MDL Number: MFCD06659909
CAS Number: 850429-61-7

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR