1-Bromo-3-(cyclopropylmethyl)thiobenzene ≥95%

Supplier: Matrix Scientific

069679-1G 069679-5G 069679-25G
101941-404EA 175.38 USD
101941-404 101941-406 101941-408
1-Bromo-3-(cyclopropylmethyl)thiobenzene ≥95%
1-Bromo-3-(cyclopropylmethyl)thiobenzene

Hazard: Irritant

Formula: C₁₀H₁₁BrS
MDL Number: MFCD09870070
CAS Number: 1000576-47-5

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR