3-Chloro-6-methylpyridazine-4-carboxylic acid ≥96%

Supplier: Matrix Scientific

069673-500MG 069673-1G 069673-5G
101941-374EA 146.15 USD
101941-374 101941-376 101941-378
3-Chloro-6-methylpyridazine-4-carboxylic acid ≥96%
3-Chloro-6-methylpyridazine-4-carboxylic acid

Hazard: Irritant

Formula: C₆H₅ClN₂O₂
MDL Number: MFCD08235214
CAS Number: 914637-40-4

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR