1-(Tetrahydropyranyl)-1H-indazole-5-carboxylic acid

Supplier: Matrix Scientific

069671-1G 069671-5G
101941-364EA 524.62 USD
101941-364 101941-366
1-(Tetrahydropyranyl)-1H-indazole-5-carboxylic acid
1-(Tetrahydropyranyl)-1H-indazole-5-carboxylic acid

Hazard: Irritant

Formula: C₁₃H₁₄N₂O₃
MW: 246.27 g/mol
MDL Number: MFCD09870060
CAS Number: 1000576-28-2

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR