1-Methyl-1H-indazole-7-boronic acid ≥95%

Supplier: Matrix Scientific

Synonyms: (1-Methyl-1H-indazol-7-yl)boronic acid

069670-500MG 069670-1G 069670-5G
101941-358EA 141.54 USD
101941-358 101941-360 101941-362
1-Methyl-1H-indazole-7-boronic acid ≥95%
1-Methyl-1H-indazole-7-boronic acid

Hazard: Irritant/Refrigerate

Formula: C₈H₉BN₂O₂
MW: 175.9803 g/mol
MDL Number: MFCD09870054
CAS Number: 1001907-59-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR