4-Bromo-2-fluoro-3-formylpyridine ≥97%

Supplier: Matrix Scientific

Synonyms: 4-Bromo-2-fluoronicotinaldehyde

069666-1G 069666-5G 069666-25G
101941-338EA 640 USD
101941-338 101941-340 101941-342
4-Bromo-2-fluoro-3-formylpyridine ≥97%
4-Bromo-2-fluoro-3-formylpyridine

Hazard: Irritant

Formula: C₆H₃BrFNO
MW: 204 g/mol
MDL Number: MFCD09870050
CAS Number: 128071-77-2

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR