7-Bromo-2-methyl-2H-indazole ≥98%

Supplier: Matrix Scientific

069664-1G 069664-5G
101941-328EA 223.08 USD
101941-328 101941-330
7-Bromo-2-methyl-2H-indazole ≥98%
7-Bromo-2-methyl-2H-indazole

Hazard: Irritant

Formula: C₈H₇BrN₂
MW: 211.600 g/mol
MDL Number: MFCD09870049
CAS Number: 701910-14-7

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR