6-Bromo-2-methyl-2H-indazole ≥97%

Supplier: Matrix Scientific

069663-1G 069663-5G
101941-324EA 815.38 USD
101941-324 101941-326
6-Bromo-2-methyl-2H-indazole ≥97%
6-Bromo-2-methyl-2H-indazole

Hazard: Irritant

Formula: C₈H₇BrN₂
MW: 211.0600 g/mol
MDL Number: MFCD09870048
CAS Number: 590417-95-1

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR