5-Bromo-2-methyl-2H-indazole ≥97%

Supplier: Matrix Scientific

Synonyms: 2-Methyl-5-bromo-2H-indazole

069662-1G 069662-5G
101941-320EA 815.38 USD
101941-320 101941-322
5-Bromo-2-methyl-2H-indazole ≥97%
5-Bromo-2-methyl-2H-indazole

Hazard: Irritant

Formula: C₈H₇BrN₂
MDL Number: MFCD09870047
CAS Number: 465529-56-0

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR