4-Bromo-2-methyl-2H-indazole ≥97%

Supplier: Matrix Scientific

069661-1G 069661-5G
101941-316EA 524.62 USD
101941-316 101941-318
4-Bromo-2-methyl-2H-indazole ≥97%
4-Bromo-2-methyl-2H-indazole

Hazard: Irritant

Formula: C₈H₇BrN₂
MW: 211.0586 g/mol
MDL Number: MFCD09870046
CAS Number: 590417-93-9

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR