1-Methyl-1H-indazole-4-boronic acid ≥97%

Supplier: Matrix Scientific

Synonyms: (1-Methyl-1H-indazol-4-yl)boronic acid

069660-500MG 069660-1G 069660-5G
101941-310EA 123.08 USD
101941-310 101941-312 101941-314
1-Methyl-1H-indazole-4-boronic acid ≥97%
1-Methyl-1H-indazole-4-boronic acid

Hazard: Irritant

Formula: C₈H₉BN₂O₂
MW: 175.9803 g/mol
MDL Number: MFCD09870045
CAS Number: 1001907-60-3

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR