7-Bromo-1-methyl-1H-indazole ≥98%
Supplier: Matrix Scientific
Synonyms:
7-Bromo-1-methylindazole
069659-1G
069659-5G
101941-306EA
246.15
USD
101941-306
101941-308
7-Bromo-1-methyl-1H-indazole ≥98%
7-Bromo-1-methyl-1H-indazole
Hazard: Irritant
Formula:
C₈H₇BrN₂ |
MDL Number:
MFCD09878570 CAS Number: 1000576-59-9 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...