5-Bromo-1-methyl-1H-indazole ≥98%

Supplier: Matrix Scientific

069656-1G 069656-5G
101941-294EA 524.62 USD
101941-294 101941-296
5-Bromo-1-methyl-1H-indazole ≥98%
5-Bromo-1-methyl-1H-indazole

Hazard: Irritant

Formula: C₈H₇BrN₂
MW: 211.0600 g/mol
MDL Number: MFCD09878568
CAS Number: 465529-57-1

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR